Newsroom Alerts
To opt-in for email alerts, please enter your email address in the field below and select at least one alert option. After submitting your request, you will receive an activation email to the requested email address. You must click the activation link in order to complete your subscription. You can sign up for additional alert options at any time.
At Thermo Fisher Scientific, we promise to treat your data with respect and will not share your information with any third party. You can unsubscribe to any of the email alerts you are subscribed to by visiting the ‘unsubscribe’ section below. If you experience any issues with this process, please contact us for further assistance.
By providing your email address below, you are providing consent to Thermo Fisher Scientific to send you the requested Email Alert updates.
* Required
News Details
View all news
Thermo Fisher Scientific Launches TrueMark STI Select Panel
02/08/2023
Research Panel Detects Four of the Most Common Sexually Transmitted Infectious Pathogens with One Panel
CARLSBAD, Calif.--(BUSINESS WIRE)-- Thermo Fisher Scientific, the world leader in serving science, today announced the launch of its Applied Biosystems TrueMark STI Select Panel, a polymerase chain reaction (PCR) research use only test designed to detect Chlamydia trachomatis, Neisseria gonorrhoeae, Trichomonas vaginalis and Mycoplasma genitalium in one test, as well as RNase P, included as a human internal control.
Cases of chlamydia, gonorrhea, and syphilis have all increased over the past two years, according to the Centers for Disease Control and Prevention (CDC)1. And, with over two and a half million cases estimated in 2021, these diseases show no signs of slowing down. These sexually transmitted infections (STIs) can often have overlapping symptoms or occur as co-infections alongside other STIs, sometimes making identifying them time-consuming. The Applied Biosystems TrueMark STI Select Panel simultaneously tests for the four common STIs on the same panel, helping labs save time that would have been otherwise needed to run each on their own. This panel will enable researchers to detect these sexually transmitted infectious microorganisms faster and with higher accuracy, and help enable understanding of testing regimens and potential treatments in the future.
“STIs are steadily increasing and testing for these pathogens is an important part of understanding how and where these diseases are spreading,” said Dr. Manoj Gandhi, senior medical director of Genetic Testing Solutions at Thermo Fisher Scientific. “The TrueMark STI Select Panel helps labs get answers quickly by consolidating four tests into a single assay, empowering them to paint a complete picture of the disease state for their research.”
The test can analyze samples collected from vaginal or genital swabs in one reaction well and is optimized for use with QuantStudio real-time PCR instruments. These samples can also be prepared using workflows that currently exist in most labs that use the Applied Biosystems MagMAX Viral/Pathogen kits automated on a KingFisher Purification System instrument and mixed with the Applied Biosystems multiplex master mix onto a 96-well or 384-well plate. The high specificity and sensitivity panel can be used by researchers to study outbreaks of STIs at a much faster rate, providing results within four hours, whereas other PCR tests for just one target can take over three hours each.
The TrueMark STI Select Panel is sold as a combo kit, which includes the STI panel assay and master mix; positive controls are also included for added accuracy and efficiency.
For more information on Thermo Fisher’s TrueMark STI Panel, please visit: https://www.thermofisher.com/truemarkstiselectpanel
For Research Use Only. Not for use in diagnostic procedures.
About Thermo Fisher
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue of approximately $40 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, increasing productivity in their laboratories, improving patient health through diagnostics or the development and manufacture of life-changing therapies, we are here to support them. Our global team delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services, Patheon and PPD. For more information, please visit www.thermofisher.com.
1 https://www.cdc.gov/std/statistics/2021/default.htm
Media:
Catie Campbell
Phone: 442-303-2104
E-mail: catie.campbell@thermofisher.com
Ariane Lovell
Phone: 646-307-6317
Email: Ariane.lovell@finnpartners.com
Source: Thermo Fisher
Multimedia Files:
View all news