Newsroom Alerts
To opt-in for email alerts, please enter your email address in the field below and select at least one alert option. After submitting your request, you will receive an activation email to the requested email address. You must click the activation link in order to complete your subscription. You can sign up for additional alert options at any time.
At Thermo Fisher Scientific, we promise to treat your data with respect and will not share your information with any third party. You can unsubscribe to any of the email alerts you are subscribed to by visiting the ‘unsubscribe’ section below. If you experience any issues with this process, please contact us for further assistance.
By providing your email address below, you are providing consent to Thermo Fisher Scientific to send you the requested Email Alert updates.
* Required
News Details
View all news
Thermo Scientific Explorer 4 Additive Scanning Electron Microscope Delivers First Dedicated Solution for 3D Printing Process Control
04/26/2018
FORT WORTH, Texas, April 26, 2018 /PRNewswire/ -- Rapid + TCT 2018 – Manufacturers using additive manufacturing (AM) processes can now easily use the high-resolution imaging and elemental analysis capabilities of scanning electron microscopy (SEM) to characterize powders that are the raw materials for many of their processes. The new Thermo Scientific Explorer 4 Additive is the first commercially-released SEM specifically designed to measure particle size, shape and composition in AM metal powders, and to inspect finished parts to assure quality. The new Explorer 4 Additive solution will be demonstrated at Rapid + TCT 2018 (booth 625) in Fort Worth, Texas, April 24-26, 2018.
"The use of additive manufacturing processes is growing rapidly in many industries, especially aerospace, automotive and medical, and the technology is quickly advancing," said Trisha Rice, vice president and general manager, materials science, Thermo Fisher Scientific. "The Explorer 4 Additive provides critical visibility into these processes, which may lead to better understanding, tighter control, improved yields and higher quality."
The Explorer 4 Additive automatically and simultaneously analyzes three of the most critical characteristics of powders used in powder-bed and powder-fed AM processes:
  • Particle size distribution – SEM can measure the entire size range of AM powders with better accuracy than competing techniques.
  • Morphology – SEM has the resolution needed to distinguish subtle differences in shape that can greatly affect the flow and packing behavior of the powder.
  • Impurities detection – Advanced energy dispersive X-ray (EDX) spectrometry provides fast, elemental analysis that can automatically identify impurities. Suspect particles can then be easily relocated for more detailed examination.
The Explorer 4 Additive system's ability to examine and classify large sets of particles, inclusions, voids and cracks within minutes enables the use of statistical process control techniques and permits faster responses to process excursions. Its high-resolution imaging and micro-analysis capabilities enable failure analysis and process engineers to quickly find the root causes of process and product failures.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue exceeding $30 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, improving patient diagnostics and therapies or increasing productivity in their laboratories, we are here to support them. Our global team of more than 80,000 colleagues delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services and Patheon. For more information, please visit www.thermofisher.com.
Media Contact Information:
Kathy Gill
Thermo Fisher Scientific
+1 503-726-7684 
kathy.gill@thermofisher.com  
Sandy Fewkes
MindWrite Communications
+1 408-224-4024
sandy@mind-write.com
SOURCE Thermo Fisher Scientific
If you have any questions regarding this Press Release, please email us at media.relations@thermofisher.com.
Multimedia Files:
View all news